Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Desmocollin-3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4630
This item is not returnable.
View return policy
Description
Desmocollin-3 Polyclonal specifically detects Desmocollin-3 in Human samples. It is validated for Western Blot.Specifications
Desmocollin-3 | |
Polyclonal | |
Unconjugated | |
Q14574 | |
DSC3 | |
Mouse myeloma cell line NS0-derived recombinant human Desmocollin-3 Arg136-Lys690 Accession # Q14574 | |
100 μg | |
Primary | |
Detects human Desmocollin-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Desmocollin-2 is observed and less than 1% cross-reactivity with rhDesmocollin-1 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Cadherin family member 3, CDHF3, CDHF3HT-CP, desmocollin 3, Desmocollin3, desmocollin-4, DSC, DSC1, DSC2, DSC3, DSC4, DSC4desmocollin-3, HT-CP | |
Sheep | |
Affinity Purified | |
RUO | |
1825 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction