Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Desmoglein-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB944
This item is not returnable.
View return policy
Description
Desmoglein-1 Monoclonal specifically detects Desmoglein-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Desmoglein-1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cadherin family member 4, CDHF4, CDHF4PPKS1, desmoglein 1, desmoglein-1, Desmoglein1, Desmosomal glycoprotein 1, DG1, DGI, DSG, DSG1, Pemphigus foliaceus antigen, SPPK1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1828 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Immunohistochemistry | |
129204 | |
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Q02413 | |
DSG1 | |
Mouse myeloma cell line NS0-derived recombinant human Desmoglein-1 Glu50-His545 Accession # Q02413 | |
500 μg | |
Primary | |
Detects human Desmoglein-1 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Desmoglein-2 or rhDesmoglein-3. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction