Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Desmoglein-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $420.50
Specifications
Antigen | Desmoglein-1 |
---|---|
Clone | 129209 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9441SP
![]() |
R&D Systems
MAB9441SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9441100
![]() |
R&D Systems
MAB9441100 |
100 μg |
Each for $420.50
|
|
|||||
Description
Desmoglein-1 Monoclonal antibody specifically detects Desmoglein-1 in Human samples. It is validated for ImmunohistochemistrySpecifications
Desmoglein-1 | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
Q02413 | |
1828 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
129209 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Cadherin family member 4, CDHF4, CDHF4PPKS1, desmoglein 1, desmoglein-1, Desmoglein1, Desmosomal glycoprotein 1, DG1, DGI, DSG, DSG1, Pemphigus foliaceus antigen, SPPK1 | |
Mouse myeloma cell line NS0-derived recombinant human Desmoglein-1, Glu50-His545, Accession # Q02413 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title