Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Desmoglein-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB947
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB947 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB947 Supplier R&D Systems Supplier No. MAB947
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Desmoglein-2 Monoclonal specifically detects Desmoglein-2 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Specifications

Antigen Desmoglein-2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 141409
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. CAA81226
Gene Alias ARVC10, ARVD10, Cadherin family member 5, CDHF5, CDHF5MGC117034, CMD1BB, desmoglein 2, desmoglein-2, Desmoglein2, DSG2, HDGC, MGC117036, MGC117037
Gene Symbols DSG2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Desmoglein-2 Ala50-Gly608 (predicted) Accession # CAA81226
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1829
Test Specificity Detects human Desmoglein-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 25-50% cross-reactivity with recombinant human (rh) Desmoglein-1 is observed and no cross-reactivity with rhDesmoglein-3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.