Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Desmoglein-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB947SP
This item is not returnable.
View return policy
Description
Desmoglein-2 Monoclonal specifically detects Desmoglein-2 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
Desmoglein-2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ARVC10, ARVD10, Cadherin family member 5, CDHF5, CDHF5MGC117034, CMD1BB, desmoglein 2, desmoglein-2, Desmoglein2, DSG2, HDGC, MGC117036, MGC117037 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1829 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunocytochemistry | |
141409 | |
Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 5-25 ug/mL | |
CAA81226 | |
DSG2 | |
Mouse myeloma cell line NS0-derived recombinant human Desmoglein-2 Ala50-Gly608 (predicted) Accession # CAA81226 | |
25 μg | |
Primary | |
Detects human Desmoglein-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 25-50% cross-reactivity with recombinant human (rh) Desmoglein-1 is observed and no cross-reactivity with rhDesmoglein-3 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction