Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Desmoglein-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF947
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF947 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF947 Supplier R&D Systems Supplier No. AF947
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

Desmoglein-2 Polyclonal specifically detects Desmoglein-2 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Desmoglein-2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Simple Western 5 ug/mL
Gene Accession No. CAA81226
Gene Alias ARVC10, ARVD10, Cadherin family member 5, CDHF5, CDHF5MGC117034, CMD1BB, desmoglein 2, desmoglein-2, Desmoglein2, DSG2, HDGC, MGC117036, MGC117037
Gene Symbols DSG2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Desmoglein-2 Met1-Gly608 Accession # CAA81226
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1829
Test Specificity Detects human Desmoglein-2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Desmoglein-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.