Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Desmoglein-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Supplier: R&D Systems MAB1720
This item is not returnable.
View return policy
Description
Desmoglein-3 Monoclonal specifically detects Desmoglein-3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Desmoglein-3 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cadherin family member 6, CDHF6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, Desmoglein3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, DSG3, pemphigus vulgaris antigen, PVA | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1830 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunohistochemistry | |
216519 | |
Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P32926 | |
DSG3 | |
Mouse myeloma cell line NS0-derived recombinant human Desmoglein-3 Glu50-Arg615 Accession # P32926 | |
500 μg | |
Primary | |
Detects human Desmoglein-3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Desmoglein-1, rhDesmoglein-2, or rhCadherin-17 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction