Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Desmoglein-4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6589
This item is not returnable.
View return policy
Description
Desmoglein-4 Polyclonal specifically detects Desmoglein-4 in Human samples. It is validated for Western Blot.Specifications
Desmoglein-4 | |
Polyclonal | |
Unconjugated | |
Q86SJ6 | |
DSG4 | |
Mouse myeloma cell line NS0-derived recombinant human Desmoglein-4 Glu24-Ala631 Accession # Q86SJ6 | |
100 μg | |
Primary | |
Detects human Desmoglein-4 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Desmoglein-1, rhDesmoglein-2, and rhDesmoglein-3 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Cad13, Cadherin family member 13, CDGF13, CDHF13, CDHF13CDH family member 13, desmoglein 4, Desmoglein4, DSG4, LAHdesmoglein-4 | |
Sheep | |
Affinity Purified | |
RUO | |
147409 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction