Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Desmoplakin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9080SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9080SP 25 μg
MAB9080 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9080SP Supplier R&D Systems Supplier No. MAB9080SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Desmoplakin Monoclonal specifically detects Desmoplakin in Human samples. It is validated for Western Blot.

Specifications

Antigen Desmoplakin
Applications Western Blot
Classification Monoclonal
Clone 824038
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P15924
Gene Alias 250/210 kDa paraneoplastic pemphigus antigen, DCWHKTA, desmoplakin, desmoplakin (DPI, DPII), desmoplakin I, desmoplakin II, DP, DPI, DPII, DSP, KPPS2, PPKS2
Gene Symbols DSP
Host Species Mouse
Immunogen E. coli-derived recombinant human Desmoplakin Asn1796-Val2046 Accession # P15924
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1832
Test Specificity Detects human Desmoplakin in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.