Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DFF40/CAD Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6289SP
This item is not returnable.
View return policy
Description
DFF40/CAD Monoclonal specifically detects DFF40/CAD in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
DFF40/CAD | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
O76075 | |
DFFB | |
E. coli-derived recombinant human DFF40/CAD Met1-Gln338 Accession # O76075 | |
25 μg | |
Primary | |
Detects human DFF40/CAD in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
609809 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CAD, CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPAN, CPANDNA fragmentation factor 40 kDa subunit, DFF2, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DFFB, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.- | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1677 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction