Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DGK-zeta Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF6099SP
This item is not returnable.
View return policy
Description
DGK-zeta Polyclonal specifically detects DGK-zeta in Human samples. It is validated for Western Blot.Specifications
DGK-zeta | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DAGK5, DAGK6DAG kinase zeta, DGKZ, DGKzeta, DGK-ZETA, diacylglycerol kinase zeta, diacylglycerol kinase, zeta, diacylglycerol kinase, zeta 104kDa, Diglyceride kinase zeta, EC 2.7.1.107, hDGKzeta | |
Goat | |
Affinity Purified | |
RUO | |
8525 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q13574 | |
DGKZ | |
E. coli-derived recombinant human DGK-zeta Gln800-Val945 Accession # Q13574 | |
25 μg | |
Primary | |
Detects human DGK-zeta in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction