Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DGK-zeta Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF6099
This item is not returnable.
View return policy
Description
DGK-zeta Polyclonal specifically detects DGK-zeta in Human samples. It is validated for Western Blot.Specifications
DGK-zeta | |
Polyclonal | |
Unconjugated | |
Q13574 | |
DGKZ | |
E. coli-derived recombinant human DGK-zeta Gln800-Val945 Accession # Q13574 | |
100 μg | |
Primary | |
Detects human DGK-zeta in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
DAGK5, DAGK6DAG kinase zeta, DGKZ, DGKzeta, DGK-ZETA, diacylglycerol kinase zeta, diacylglycerol kinase, zeta, diacylglycerol kinase, zeta 104kDa, Diglyceride kinase zeta, EC 2.7.1.107, hDGKzeta | |
Goat | |
Affinity Purified | |
RUO | |
8525 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction