Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Digoxigenin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | Digoxigenin |
---|---|
Clone | 2577B |
Dilution | Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 1-25 ug/mL |
Applications | Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10386SP
![]() |
R&D Systems
MAB10386SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10386100
![]() |
R&D Systems
MAB10386100 |
100 μg |
Each for $479.50
|
|
|||||
Description
Digoxigenin Monoclonal specifically detects Digoxigenin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
Digoxigenin | |
Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 1-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
KLH-coupled Digoxigenin | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2577B | |
Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
DIG | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title