Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DISP1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3549SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3549SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3549SP Supplier R&D Systems Supplier No. AF3549SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

DISP1 Polyclonal specifically detects DISP1 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen DISP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_116279
Gene Alias DISPA, DISPAFLJ43740, Dispatched, dispatched A, dispatched homolog 1 (Drosophila), DKFZp434I0428, MGC104180, MGC13130, MGC16796, protein dispatched homolog 1
Gene Symbols DISP1
Host Species Sheep
Immunogen E. coli-derived recombinant human DISP1 Glu212-Lys492, Ser746-Thr984 Accession # NP_116279
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84976
Test Specificity Detects human DISP1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.