Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DISP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3549
This item is not returnable.
View return policy
Description
DISP1 Monoclonal specifically detects DISP1 in Human samples. It is validated for Immunocytochemistry.Specifications
DISP1 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q96F81 | |
DISP1 | |
E. coli-derived recombinant human DISP1 Glu212-Lys492 and Ser746-Thr984 Accession # Q96F81 | |
100 μg | |
Primary | |
Detects human DISP1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) DISP2 or rhSMAGP is observed. | |
Human | |
Purified |
Immunocytochemistry | |
768213 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DISPA, DISPAFLJ43740, Dispatched, dispatched A, dispatched homolog 1 (Drosophila), DKFZp434I0428, MGC104180, MGC13130, MGC16796, protein dispatched homolog 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
84976 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction