Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DISP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3549SP
This item is not returnable.
View return policy
Description
DISP1 Monoclonal specifically detects DISP1 in Human samples. It is validated for Immunocytochemistry.Specifications
DISP1 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q96F81 | |
DISP1 | |
E. coli-derived recombinant human DISP1 Glu212-Lys492 and Ser746-Thr984 Accession # Q96F81 | |
25 μg | |
Primary | |
Detects human DISP1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) DISP2 or rhSMAGP is observed. | |
Human | |
Purified |
Immunocytochemistry | |
768213 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DISPA, DISPAFLJ43740, Dispatched, dispatched A, dispatched homolog 1 (Drosophila), DKFZp434I0428, MGC104180, MGC13130, MGC16796, protein dispatched homolog 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
84976 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction