Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dkk-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$200.50 - $655.00
Specifications
| Antigen | Dkk-1 |
|---|---|
| Clone | 141119 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1096SP
![]() |
R&D Systems
MAB1096SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1096050
![]() |
R&D Systems
MAB1096050 |
50 μg |
Each for $292.00
|
|
|||||
MAB1096500
![]() |
R&D Systems
MAB1096500 |
500 μg |
Each for $655.00
|
|
|||||
Description
Dkk-1 Monoclonal specifically detects Dkk-1 in Human samples. It is validated for Western Blot.Specifications
| Dkk-1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O94907 | |
| 22943 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Dkk-1 Thr32-His266 Accession # O94907 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Dkk-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 50% cross-reactivity with recombinant mouse (rm) Dkk-1 and no cross-reactivity with rmDkk-2, rhDkk-3, or rhDkk-4. |
| 141119 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, Dkk1, DKK-1, hDkk-1, SKdickkopf-1 like | |
| DKK1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title