Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Dkk-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6628SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6628SP 25 μg
MAB6628100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6628SP Supplier R&D Systems Supplier No. MAB6628SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dkk-2 Monoclonal specifically detects Dkk-2 in Human samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Dkk-2
Applications Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 994930
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 μg/mL, Intracellular Staining by Flow Cytometry 0.25 μg/106 cells, Immunocytochemistry 5-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. NP_055236
Gene Alias dickkopf (Xenopus laevis) homolog 2, dickkopf 2, dickkopf homolog 2 (Xenopus laevis), dickkopf related protein-2, Dickkopf-2, dickkopf-related protein 2, Dkk2, DKK-2, hDkk-2
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human Dkk-2, Met1-Ile259, Accession # NP_055236
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 27123
Test Specificity Detects human Dkk-2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.