Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dkk-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | Dkk-3 |
---|---|
Clone | 138728 |
Concentration | LYOPH |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11181SP
![]() |
R&D Systems
MAB11181SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11181
![]() |
R&D Systems
MAB11181 |
500 μg |
Each for $667.00
|
|
|||||
Description
Dkk-3 Monoclonal specifically detects Dkk-3 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
Dkk-3 | |
LYOPH | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q9UBP4.1 | |
27122 | |
Mouse myeloma cell line NS0-derived recombinant human Dkk-3 Ala22-Ile350 Accession # Q9UBP4.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Dkk-3 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Dkk-1, recombinant mouse (rm) Dkk-2, or rmDkk-3 is observed. |
138728 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk3, Dkk-3, hDkk-3, regulated in glioma, REIC, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
DKK3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title