Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dkk-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Supplier: R&D Systems AF1118
This item is not returnable.
View return policy
Description
Dkk-3 Polyclonal specifically detects Dkk-3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Dkk-3 | |
Polyclonal | |
Unconjugated | |
Q9UBP4 | |
DKK3 | |
Mouse myeloma cell line NS0-derived recombinant human Dkk-3 Ala22-Ile350 Accession # Q9UBP4 | |
100 μg | |
Primary | |
Detects human Dkk-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant mouse Dkk-3 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk3, Dkk-3, hDkk-3, regulated in glioma, REIC, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
Goat | |
Affinity Purified | |
RUO | |
27122 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction