Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dkk-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $2350.00
Specifications
Antigen | Dkk-4 |
---|---|
Clone | 185709 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1269SP
![]() |
R&D Systems
MAB1269SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1269050
![]() |
R&D Systems
MAB1269050 |
50 μg |
Each for $292.00
|
|
|||||
MAB1269500
![]() |
R&D Systems
MAB1269500 |
500 μg |
Each for $2,350.00
|
|
|||||
Description
Dkk-4 Monoclonal specifically detects Dkk-4 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
Dkk-4 | |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q9UBT3 | |
27121 | |
Mouse myeloma cell line NS0-derived recombinant human Dkk-4 Leu19-Leu224 Accession # Q9UBT3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Dkk-4 in ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse (rm) Dkk-4 is observed and no cross-reactivity with recombinant human (rh) Dkk-1 or rhDkk-3 is observed. In sandwich immunoassays, less than 2% cross-reactivity with rmDkk-4 is observed and no cross-reactivity with rhDkk-1, recombinant rat Dkk-1, rmDkk-1, or rhDkk-3 is observed. |
185709 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dickkopf (Xenopus laevis) homolog 4, dickkopf homolog 4 (Xenopus laevis), dickkopf-4, dickkopf-related protein 4, Dkk4, DKK-4, hDkk-4, MGC129562, MGC129563 | |
DKK4 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title