Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DLEC/CLEC4C/BDCA-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Specifications
Antigen | DLEC/CLEC4C/BDCA-2 |
---|---|
Clone | 992258 |
Dilution | Immunohistochemistry 1-25μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB62991-SP
![]() |
R&D Systems
MAB62991SP |
25 μg |
Each for $214.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
MAB62991-100
![]() |
R&D Systems
MAB62991100 |
100 μg |
Each for $467.50
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
DLEC/CLEC4C/BDCA-2 Monoclonal specifically detects DLEC/CLEC4C/BDCA-2 in Human samples. It is validated for Immunohistochemistry.Specifications
DLEC/CLEC4C/BDCA-2 | |
Immunohistochemistry 1-25μg/mL | |
Monoclonal | |
Supernatant | |
Human | |
Q8WTT0 | |
170482 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
992258 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
BDCA2, BDCA-2, BDCA2MGC125791, Blood dendritic cell antigen 2, blood dendritic cell antigen 2 protein, CD303, CD303 antigen, CLEC4C, CLECSF11, CLECSF11PRO34150, CLECSF7, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 11, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 7, C-type lectin domain family 4 member C, C-type lectin superfamily member 7, dendritic cell lectin b, Dendritic lectin, DLEC, DLECMGC125792, HECL, HECLMGC125793, MGC125789, PRO34150 | |
CHO Chinese hamster ovary cell line transfected with human DLEC/CLEC4C/BDCA-2 Phe46-Ile213 Accession # Q8WTT0 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human DLEC/CLEC4C/BDCA-2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title