Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DLEC/CLEC4C/BDCA-2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1376
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1376 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1376 Supplier R&D Systems Supplier No. BAF1376
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

DLEC/CLEC4C/BDCA-2 Polyclonal specifically detects DLEC/CLEC4C/BDCA-2 in Human samples. It is validated for Western Blot.

Specifications

Antigen DLEC/CLEC4C/BDCA-2
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. Q8WTT0
Gene Alias BDCA2, BDCA-2, BDCA2MGC125791, Blood dendritic cell antigen 2, blood dendritic cell antigen 2 protein, CD303, CD303 antigen, CLEC4C, CLECSF11, CLECSF11PRO34150, CLECSF7, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 11, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 7, C-type lectin domain family 4 member C, C-type lectin superfamily member 7, dendritic cell lectin b, Dendritic lectin, DLEC, DLECMGC125792, HECL, HECLMGC125793, MGC125789, PRO34150
Gene Symbols CLEC4C
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human DLEC/CLEC4C/BDCA-2 Phe46-Ile213 Accession # Q8WTT0
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170482
Test Specificity Detects human DLEC/CLEC4C/BDCA-2 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile Tris-buffered saline, pH 7.3 (20 mM Trizma base, 150 mM NaCl) containing 0.1% bovine serum albumin.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.