Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DLL3 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$212.05 - $615.21

Specifications

Antigen DLL3
Clone 378703
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB4315SP
GREENER_CHOICE
View Documents
R&D Systems
MAB4315SP
25 μg
Each for $212.05
Only null left
Add to Cart
 
MAB4315
GREENER_CHOICE
View Documents
R&D Systems
MAB4315
100 μg
Each for $615.21
Only null left
Add to Cart
 
Description

Description

DLL3 Monoclonal specifically detects DLL3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

DLL3
LYOPH
Western Blot
Unconjugated
Mouse
Human
Q9NYJ7
10683
Chinese hamster ovary cell line CHO-derived recombinant human DLL3 Ala27-Pro488 Accession # Q9NYJ7
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human DLL3 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) DLL1 or rhDLL4.
378703
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, Pudgy, SCDO1, SCDO1delta3
DLL3
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.