Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DLL3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB4315
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB4315 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB4315 Supplier R&D Systems Supplier No. AB4315
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

DLL3 Polyclonal specifically detects DLL3 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen DLL3
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells
Gene Accession No. Q9NYJ7
Gene Alias delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, Pudgy, SCDO1, SCDO1delta3
Gene Symbols DLL3
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human DLL3 Ala27-Pro488 Accession # Q9NYJ7
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10683
Test Specificity Detects human DLL3 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) DLL1 is observed, and less than 8% cross-reactivity with rhDLL4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.