Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DLL3 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4315P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4315P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4315P Supplier R&D Systems Supplier No. FAB4315P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

DLL3 Polyclonal specifically detects DLL3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen DLL3
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q9NYJ7
Gene Alias delta (Drosophila)-like 3, Delta3, delta-like 3 (Drosophila), delta-like protein 3, Drosophila Delta homolog 3, Pudgy, SCDO1, SCDO1delta3
Gene Symbols DLL3
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human DLL3 Ala27-Pro488 Accession # Q9NYJ7
Purification Method Protein A or G purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10683
Test Specificity Detects human DLL3 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) DLL1 is observed and less than 8% cross-reactivity with rhDLL4 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.