Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DLL4 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB1506R100UG
This item is not returnable.
View return policy
Description
DLL4 Monoclonal specifically detects DLL4 in Human samples. It is validated for Flow Cytometry.Specifications
DLL4 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human DLL4 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Flow Cytometry | |
447506 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q9NR61 | |
DLL4 | |
Mouse myeloma cell line NS0-derived recombinant human DLL4, Ser27-Pro524, Accession # Q9NR61 | |
100 μg | |
54567 | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction