Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DMP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
| Antigen | DMP-1 |
|---|---|
| Clone | 560716 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
Description
DMP-1 Monoclonal specifically detects DMP-1 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| DMP-1 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARHP, ARHR, dentin matrix acidic phosphoprotein, dentin matrix acidic phosphoprotein 1, Dentin matrix protein 1, DMP1, DMP-1 | |
| DMP1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 560716 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_001073380 | |
| 1758 | |
| Mouse myeloma cell line NS0-derived recombinant human DMP-1 isoform 2 Leu17-Tyr497 Accession # NP_001073380 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects human DMP-1 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse DMP-1, recombinant human (rh) CD44, and rhComplement Factor H is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title