Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DMP-1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM41291
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM41291 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM41291 Supplier R&D Systems Supplier No. BAM41291
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DMP-1 Monoclonal specifically detects DMP-1 in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).

Specifications

Antigen DMP-1
Applications ELISA
Classification Monoclonal
Clone 560722
Concentration LYOPH
Conjugate Biotin
Dilution ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. NP_001073380
Gene Alias ARHP, ARHR, dentin matrix acidic phosphoprotein, dentin matrix acidic phosphoprotein 1, Dentin matrix protein 1, DMP1, DMP-1
Gene Symbols DMP1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human DMP-1 isoform 2 Leu17-Tyr497 Accession # NP_001073380
Purification Method Protein A or G purified from ascites
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1758
Test Specificity Detects human DMP-1 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse DMP-1, recombinant human (rh) CD44, and rhComplement Factor H is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.