Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DNAM-1/CD226 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB666T100UG
This item is not returnable.
View return policy
Description
DNAM-1/CD226 Monoclonal specifically detects DNAM-1/CD226 in Human samples. It is validated for Flow Cytometry.Specifications
DNAM-1/CD226 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q15762 | |
CD226 | |
Mouse myeloma cell line NS0-derived recombinant human DNAM-1/CD226 Glu19-Asn247 (predicted) Accession # Q15762 | |
RUO | |
10666 | |
Human | |
IgG1 |
Flow Cytometry | |
102511 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD226, CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
Mouse | |
100 μg | |
Primary | |
Detects human DNAM-1/CD226 in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction