Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DNAM-1/CD226 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6661100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6661100 100 μg
MAB6661SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6661100 Supplier R&D Systems Supplier No. MAB6661100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNAM-1/CD226 Monoclonal antibody specifically detects DNAM-1/CD226 in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen DNAM-1/CD226
Applications Immunohistochemistry
Classification Monoclonal
Clone 1035512
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q15762
Gene Alias CD226, CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived human DNAM-1/CD226 protein, Glu19-Asn247, Accession # Q15762
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10666
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.