Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DNMT3A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6315SP
This item is not returnable.
View return policy
Description
DNMT3A Monoclonal specifically detects DNMT3A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
DNMT3A | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q9Y6K1 | |
DNMT3A | |
E. coli-derived recombinant human DNMT3A Ala353-Lys486 Accession # Q9Y6K1 | |
25 μg | |
Primary | |
Detects human DNMT3A in direct ELISAs. In direct ELISAS, no cross-reactivity with recombinant human DNMT1 is observed. | |
Human | |
Purified |
Immunohistochemistry, Immunocytochemistry | |
681630 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1788 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction