Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DOCK3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7134SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7134SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7134SP Supplier R&D Systems Supplier No. AF7134SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

DOCK3 Polyclonal specifically detects DOCK3 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen DOCK3
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8IZD9
Gene Alias dedicator of cytokinesis 3, KIAA0299, MOCA, modifier of cell adhesion
Gene Symbols DOCK3
Host Species Sheep
Immunogen E. coli-derived recombinant human DOCK3 Gly418-Thr656 Accession # Q8IZD9
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1795
Test Specificity Detects human DOCK3 in direct ELISAs. In direct ELISAs, less than 3% cross-reactivity with recombinant human (rh) DOCK1, rhDOCK2, and rhDOCK5 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.