Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Dopamine D1R/DRD1 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8276S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8276S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8276S100 Supplier R&D Systems Supplier No. FAB8276S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dopamine D1R/DRD1 Monoclonal specifically detects Dopamine D1R/DRD1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Dopamine D1R/DRD1
Applications Flow Cytometry
Classification Monoclonal
Clone 887438
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P21728
Gene Alias D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 R, Dopamine D1 receptor, dopamine receptor D1, DRD1, DRD1A
Gene Symbols DRD1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Dopamine D1 R/DRD1 Met1-Thr446 Accession # P21728
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1812
Test Specificity Stains human Dopamine D1 R/DRD1 transfected cells but not irrelevant transfectants in flow cytometry.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.