Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dopamine D1R/DRD1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8276SP
This item is not returnable.
View return policy
Description
Dopamine D1R/DRD1 Monoclonal specifically detects Dopamine D1R/DRD1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
Dopamine D1R/DRD1 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
P21728 | |
DRD1 | |
NS0 mouse myeloma cell line transfected with human Dopamine D1 R/DRD1 Met1-Thr446 Accession # P21728 | |
25 μg | |
Primary | |
Stains human Dopamine D1 R/DRD1 transfected cells but not irrelevant transfectants in flow cytometry. | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
887438 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
D(1A) dopamine receptor, D1DR, DADR, Dopamine D1 R, Dopamine D1 receptor, dopamine receptor D1, DRD1, DRD1A | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1812 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction