Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Dopamine D2 R/DRD2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9266SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9266SP 25 μg
MAB9266100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9266SP Supplier R&D Systems Supplier No. MAB9266SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dopamine D2R/DRD2 Monoclonal specifically detects Dopamine D2R/DRD2 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Dopamine D2R/DRD2
Applications Immunohistochemistry
Classification Monoclonal
Clone 960710
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P14416
Gene Alias D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, Dopamine D2R, dopamine receptor D2, dopamine receptor D2 isoform, DRD2, seven transmembrane helix receptor
Host Species Mouse
Immunogen Human DRD2 synthetic peptide Accession # P14416
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1813
Test Specificity Detects human Dopamine D2 R/DRD2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.