Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dopamine D5R/DRD5 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB82861S100UG
This item is not returnable.
View return policy
Description
Dopamine D5R/DRD5 Monoclonal specifically detects Dopamine D5R/DRD5 in Human samples. It is validated for Flow Cytometry.Specifications
Dopamine D5R/DRD5 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P21918 | |
DRD5 | |
NS0 mouse myeloma cell line transfected with human Dopamine D5 R/DRD5 Met1-His477 Accession # P21918 | |
RUO | |
1816 | |
Human | |
IgG2a |
Flow Cytometry | |
889022 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, DBDR, Dopamine D5 R, Dopamine D5 receptor, dopamine receptor D1B, dopamine receptor D5, DRD1B, DRD1BDBDR, DRD1L2, DRD1L2MGC10601, DRD5 | |
Mouse | |
100 μg | |
Primary | |
Detects HEK293 human embryonic kidney cell line transfected with human Dopamine D5 R/DRD5 by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction