Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DOT1L Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6546SP
This item is not returnable.
View return policy
Description
DOT1L Monoclonal specifically detects DOT1L in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation.Specifications
DOT1L | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL | |
Q8TEK3 | |
DOT1L | |
E. coli-derived recombinant human DOT1L Gly2-Lys416 Accession # Q8TEK3 | |
25 μg | |
Primary | |
Detects human DOT1L in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) PCDH-8, rhNME-3, or rhWISP-3 is observed. | |
Human | |
Purified |
Immunohistochemistry, Immunoprecipitation | |
653613 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DOT1-like protein, DOT1-like, histone H3 methyltransferase (S. cerevisiae), EC 2.1.1.43, H3-K79-HMTase, KIAA1814DKFZp586P1823, KMT4, KMT4H3 lysine-79 specific, Lysine N-methyltransferase 4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
84444 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction