Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DOT1L Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6546
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6546 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6546 Supplier R&D Systems Supplier No. MAB6546
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DOT1L Monoclonal specifically detects DOT1L in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen DOT1L
Applications Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 653613
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8TEK3
Gene Alias DOT1-like protein, DOT1-like, histone H3 methyltransferase (S. cerevisiae), EC 2.1.1.43, H3-K79-HMTase, KIAA1814DKFZp586P1823, KMT4, KMT4H3 lysine-79 specific, Lysine N-methyltransferase 4
Gene Symbols DOT1L
Host Species Mouse
Immunogen E. coli-derived recombinant human DOT1L Gly2-Lys416 Accession # Q8TEK3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84444
Test Specificity Detects human DOT1L in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) PCDH-8, rhNME-3, or rhWISP-3 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.