Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DPCR-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3347
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3347 100 μg
MAB3347SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3347 Supplier R&D Systems Supplier No. MAB3347
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DPCR-1 Monoclonal specifically detects DPCR-1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen DPCR-1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 213201
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH69477
Gene Alias bCX105N19.6, C6orf37, diffuse panbronchiolitis critical region 1, DPCR1, MGC126710, MGC126712, PBLT, PBLTdiffuse panbronchiolitis critical region 1 protein
Gene Symbols DPCR1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human DPCR-1 Met1-Arg235 Accession # AAH69477
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135656
Test Specificity Detects human human DPCR-1. Stains human DPCR-1-transfected cells but not irrelevant transfectants.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.