Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DPP10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4100SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4100SP 25 μg
AF4100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4100SP Supplier R&D Systems Supplier No. AF4100SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

DPP10 Polyclonal specifically detects DPP10 in Human samples. It is validated for Western Blot.

Specifications

Antigen DPP10
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_065919
Gene Alias dipeptidyl peptidase 10, Dipeptidyl peptidase IV-related protein 3, dipeptidyl peptidase like protein 2, Dipeptidyl peptidase X, Dipeptidyl peptidase-like protein 2, dipeptidyl-peptidase 10, dipeptidyl-peptidase 10 (non-functional), DPL2, DPP X, DPPY, DPRP3, DPRP-3, DPRP3dipeptidylpeptidase 10, inactive dipeptidyl peptidase 10, KIAA1492
Gene Symbols DPP10
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human DPP10 Leu56-Glu796 (Pro340Ala) Accession # NP_065919
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57628
Test Specificity Detects human DPP10 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) DPP6 and rhDPP4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.