Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DPP3 Antibody, R&D Systems™

Rat Monoclonal Antibody
$164.60 - $437.75
Specifications
| Antigen | DPP3 |
|---|---|
| Clone | 986603 |
| Dilution | Western Blot 2 μ/mL, Immunohistochemistry 5-25 μ/mL, Immunocytochemistry 8-25 μ/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB80872SP
![]() |
R&D Systems
MAB80872SP |
25 μg |
Each for $164.60
|
|
|||||
MAB80872100
![]() |
R&D Systems
MAB80872100 |
100 μg |
Each for $437.75
|
|
|||||
Description
DPP3 Monoclonal specifically detects DPP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| DPP3 | |
| Western Blot 2 μ/mL, Immunohistochemistry 5-25 μ/mL, Immunocytochemistry 8-25 μ/mL | |
| Monoclonal | |
| Rat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Dipeptidyl aminopeptidase III, Dipeptidyl arylamidase III, dipeptidyl peptidase 3, Dipeptidyl peptidase III, dipeptidylpeptidase 3, dipeptidyl-peptidase 3, dipeptidylpeptidase III, DPP III, DPPIII, EC 3.4.14.4, FLJ11387, FLJ22331 | |
| E. coli-derived recombinant human DPP3, Ala2-Ala737, Accession No. Q9NY33 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 986603 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Human | |
| Q9NY33 | |
| 10072 | |
| IgG2b | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human DPP3 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title