Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DPP3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB80871SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB80871SP 25 μg
MAB80871100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB80871SP Supplier R&D Systems Supplier No. MAB80871SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DPP3 Monoclonal specifically detects DPP3 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen DPP3
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 987030
Conjugate Unconjugated
Dilution Western Blot 1 μg;/mL, Immunocytochemistry 8-25 μg;/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q9NY33
Gene Alias Dipeptidyl aminopeptidase III, Dipeptidyl arylamidase III, dipeptidyl peptidase 3, Dipeptidyl peptidase III, dipeptidylpeptidase 3, dipeptidyl-peptidase 3, dipeptidylpeptidase III, DPP III, DPPIII, EC 3.4.14.4, FLJ11387, FLJ22331
Host Species Mouse
Immunogen E. coli-derived recombinant human DPP3, Ala2-Ala737, Accession No. Q9NY33
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 10072.0
Test Specificity Detects human DPP3 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.