Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DPP6 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2360
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2360 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2360 Supplier R&D Systems Supplier No. BAF2360
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

DPP6 Polyclonal specifically detects DPP6 in Human samples. It is validated for Western Blot.

Specifications

Antigen DPP6
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. AAA35761
Gene Alias dipeptidyl aminopeptidase IV-related protein, dipeptidyl aminopeptidase-like protein 6, Dipeptidyl aminopeptidase-related protein, Dipeptidyl peptidase 6, Dipeptidyl peptidase IV-like protein, dipeptidyl peptidase IV-related protein, Dipeptidyl peptidase VI, dipeptidylpeptidase 6, dipeptidyl-peptidase 6, dipeptidylpeptidase VI, DPP VI, DPPX, DPPXFLJ55680, MGC46605, VF2
Gene Symbols DPP6
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human DPP6 Thr56-Asp803 (Glu458Val) Accession # AAA35761
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1804
Test Specificity Detects human DPP6 in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant mouse CD69 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.