Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DPP9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | DPP9 |
---|---|
Clone | 988515 |
Dilution | Western Blot 1μg/mL, Simple Western 10μg/mL, Immunocytochemistry 8-25μg/mL |
Applications | Immunocytochemistry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB54191SP
![]() |
R&D Systems
MAB54191SP |
25 μg |
Each for $200.50
|
|
|||||
MAB54191100
![]() |
R&D Systems
MAB54191100 |
100 μg |
Each for $514.50
|
|
|||||
Description
DPP9 Monoclonal specifically detects DPP9 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
DPP9 | |
Western Blot 1μg/mL, Simple Western 10μg/mL, Immunocytochemistry 8-25μg/mL | |
Monoclonal | |
Supernatant | |
Human | |
Q86TI2 | |
91039 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
988515 | |
Immunocytochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
dipeptidyl peptidase 9, Dipeptidyl peptidase IV-related protein 2, dipeptidyl peptidase IV-related protein-2, Dipeptidyl peptidase IX, Dipeptidyl peptidase-like protein 9, dipeptidylpeptidase 9, dipeptidyl-peptidase 9, DKFZp762F117, DP9, DPLP9, DPP IX, DPRP2, DPRP-2, EC 3.4.14.5, FLJ16073 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human DPP9 Arg2-Leu892 Accession # Q86TI2 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human DPP9 in ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title