Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DPPA4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3674SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3674SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3674SP Supplier R&D Systems Supplier No. AF3674SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

DPPA4 Polyclonal specifically detects DPPA4 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen DPPA4
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q7L190
Gene Alias developmental pluripotency associated 4, developmental pluripotency-associated protein 4, FLJ10713,2410091M23Rik
Gene Symbols DPPA4
Host Species Sheep
Immunogen E. coli-derived recombinant human DPPA4 Met1-Glu304 Accession # Q7L190
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55211
Test Specificity Detects human DPPA4 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse DPPA4 is observed and less than 1% cross-reactivity with recombinant human DPPA5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.