Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DPPIV/CD26 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1180100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1180100 100 μg
MAB1180SP 25 μg
MAB1180500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1180100 Supplier R&D Systems Supplier No. MAB1180100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen DPPIV/CD26
Applications Western Blot, Flow Cytometry, CyTOF, ELISA
Classification Monoclonal
Clone 222113
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q53TN1
Gene Alias ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPP4, DPPIV, EC 3.4.14.5
Gene Symbols DPP4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human DPPIV/CD26 Asp34-Pro766 Accession # Q53TN1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1803
Test Specificity Detects human DPPIV/CD26 in ELISAs and Western blots. In ELISAs and Western blots, no cross-reactivity with recombinant human Cathepsin A or recombinant mouse DPPIV is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.