Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DR3/TNFRSF25 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF943
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF943 100 μg
AF943SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF943 Supplier R&D Systems Supplier No. AF943
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

DR3/TNFRSF25 Polyclonal specifically detects DR3/TNFRSF25 in Human samples. It is validated for Western Blot.

Specifications

Antigen DR3/TNFRSF25
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q93038
Gene Alias Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), LARD, Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, TNFRSF25, TRAMP, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1, WSL-1
Gene Symbols TNFRSF25
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human DR3/TNFRSF25 Gln25-Phe201 Accession # Q93038
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8718
Test Specificity Detects human DR3/TNFRSF25 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse DR3 is observed, and less than 1% cross-reactivity with recombinant human (rh) Fas, rhTNF RI, rhDR6, rh4-1BB, rhCD27, rhCD30, rhCD40, rhGITR, rhHVEM, rhTNF RII and rhOPG is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.