Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DSPG3 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | DSPG3/EPYC |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
DSPG3/EPYC Polyclonal specifically detects DSPG3/EPYC in Human samples. It is validated for Western Blot.Specifications
DSPG3/EPYC | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Dermatan sulfate proteoglycan 3dermatan sulphate proteoglycan 3, DSPG3proteoglycan-lb, epiphycan, EPYC, PG-Lb, PGLB, Proteoglycan-Lb, SLRR3B, SLRR3Bepiphycan proteoglycan, Small chondroitin/dermatan sulfate proteoglycan | |
EPYC | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q99645 | |
1833 | |
Mouse myeloma cell line NS0-derived recombinant human DSPG3 Met1-Val322 Accession # Q99645 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human DSPG3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse DSPG3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title