Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DTX1/DTX4 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC7157S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC7157S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC7157S100 Supplier R&D Systems Supplier No. IC7157S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DTX1/DTX4 Monoclonal specifically detects DTX1/DTX4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen DTX1/DTX4
Applications Flow Cytometry
Classification Monoclonal
Clone 730612
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q86Y01
Gene Symbols DTX1
Host Species Mouse
Immunogen E. coli-derived recombinant human DTX1/DTX4 Met1-Phe147 Accession # Q86Y01
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human DTX1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human DTX4 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.